5,7-Dichloro-1H-indole 98%

Supplier: Ambeed
Warning

A643905-100MG A643905-250MG A643905-5G A643905-1G
76980-746EA 31.83 USD
76980-746 76980-748 76980-750 77673-748
5,7-Dichloro-1H-indole 98%
5,7-Dichloro-1H-indole

Precaución: for Research Use Only

Formula: C₈H₅Cl₂N
MW: 186.03 g/mol
MDL Number: MFCD03094982
CAS Number: 4792-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR