5,7-Dichloro-1H-indole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A188753-100MG A188753-1G A188753-250MG A188753-5G
76974-776EA 9.55 USD
76974-776 76974-778 76974-780 76974-782
5,7-Dichloro-1H-indole-2-carboxylic acid 97%
5,7-Dichloro-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅Cl₂NO₂
MW: 230.05 g/mol
MDL Number: MFCD02664420
CAS Number: 4792-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR