(2-Methoxy-5-nitrophenyl)boronic acid 98%

Supplier: Ambeed
Warning

A167871-250MG A167871-5G A167871-1G
76973-874EA 75.99 USD
76973-874 77881-300 77881-299
(2-Methoxy-5-nitrophenyl)boronic acid 98%
(2-Methoxy-5-nitrophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₈BNO₅
MDL Number: MFCD10696662
CAS Number: 677746-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR