4,4,5,5-Tetramethyl-2-(4-methyl-3-nitrophenyl)-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-3-nitrophenylboronic acid pinacol ester

A674494-1G A674494-250MG A674494-5G
76980-936EA 67 USD
76980-936 76980-938 76980-940
4,4,5,5-Tetramethyl-2-(4-methyl-3-nitrophenyl)-1,3,2-dioxaborolane 98%
4,4,5,5-Tetramethyl-2-(4-methyl-3-nitrophenyl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₈BNO₄
MDL Number: MFCD09027080
CAS Number: 1072945-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR