4-Methoxy-2-nitrophenylboronic acid pinacol ester 95%

Supplier: Ambeed
Danger

Synonyms: 2-(4-Methoxy-2-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A827611-100MG A827611-1G A827611-250MG A827611-5G
76981-750EA 34 USD
76981-750 76981-752 76981-754 76981-756
4-Methoxy-2-nitrophenylboronic acid pinacol ester 95%
4-Methoxy-2-nitrophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BNO₅
MDL Number: MFCD10699701
CAS Number: 1073353-81-7
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR