(R)-3-(Boc-amino)-3-(4-chlorophenyl)propionic acid 98%

Supplier: Ambeed
Warning

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-3-(4-chlorophenyl)propanoic acid

A245466-1G A245466-250MG A245466-5G
76976-574EA 95.49 USD
76976-574 76976-576 76976-578
(R)-3-(Boc-amino)-3-(4-chlorophenyl)propionic acid 98%
(R)-3-(Boc-amino)-3-(4-chlorophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₈ClNO₄
MW: 299.7500 g/mol
MDL Number: MFCD03427960
CAS Number: 479064-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR