3-Chloro-5-iodobenzotrifluoride 95%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-5-iodo-α,α,α-trifluorotoluene, 1-Chloro-3-iodo-5-(trifluoromethyl)benzene

A656746-10G A656746-250MG A656746-5G
76980-850EA 63.66 USD
76980-850 76980-852 77573-114
3-Chloro-5-iodobenzotrifluoride 95%
3-Chloro-5-iodobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃ClF₃I
MW: 306.451 g/mol
MDL Number: MFCD19707625
CAS Number: 1189352-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR