(S)-2-Methyl-1-phenylpropan-1-amine hydrochloride 97%

Supplier: Ambeed
Warning

A119388-100MG A119388-250MG
76971-418EA 35.01 USD
76971-418 76971-420
(S)-2-Methyl-1-phenylpropan-1-amine hydrochloride 97%
(S)-2-Methyl-1-phenylpropan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₆ClN
MW: 185.69 g/mol
MDL Number: MFCD12068472
CAS Number: 68906-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR