tert-Butyl-4-hydroxyazepane-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-hydroxyazepane-1-carboxylate, 1-Boc-4-Hydroxyazepane

A122013-100G A122013-10G A122013-1G A122013-250MG A122013-25G A122013-5G
76971-586EA 1170.18 USD
76971-586 76971-588 76971-590 76971-592 76971-594 76971-596
tert-Butyl-4-hydroxyazepane-1-carboxylate 97%
tert-Butyl-4-hydroxyazepane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
MW: 215.2900 g/mol
MDL Number: MFCD07368610
CAS Number: 478832-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR