(2S,5R)-1-Boc-2,5-dimethylpiperazine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (2S,5R)-tert-Butyl-2,5-dimethylpiperazine-1-carboxylate hydrochloride

A401774-100MG A401774-1G A401774-250MG
76979-072EA 23 USD
76979-072 76979-074 76979-076
(2S,5R)-1-Boc-2,5-dimethylpiperazine hydrochloride 95%
(2S,5R)-1-Boc-2,5-dimethylpiperazine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₂₃ClN₂O₂
MDL Number: MFCD13191717
CAS Number: 1374975-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR