(2S,4S)-1-Benzyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A335584-1G A335584-250MG A335584-25G A335584-5G
76978-334EA 119.89 USD
76978-334 76978-336 76978-338 76978-340
(2S,4S)-1-Benzyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride 95%
(2S,4S)-1-Benzyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₁₉ClN₂O₄
MW: 314.76 g/mol
MDL Number: MFCD06804628
CAS Number: 1515919-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR