1-(5-Chloropyrazin-2-yl)ethanone 95%

Supplier: Ambeed
Warning

A172036-10G A172036-1G A172036-250MG A172036-25G A172036-5G A172036-100G
76974-046EA 148 USD
76974-046 76974-048 76974-050 76974-052 77611-674 77671-446
1-(5-Chloropyrazin-2-yl)ethanone 95%
1-(5-Chloropyrazin-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O
MDL Number: MFCD12400924
CAS Number: 160252-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR