2-N-Cbz-Butane-1,2-diamine hydrochloride 95%

Supplier: Ambeed
Warning

A149209-100MG A149209-250MG
76973-036EA 66.84 USD
76973-036 76973-038
2-N-Cbz-Butane-1,2-diamine hydrochloride 95%
2-N-Cbz-Butane-1,2-diamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₉ClN₂O₂
MW: 258.74 g/mol
MDL Number: MFCD11112244
CAS Number: 1179361-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR