tert-Butyl (4-amino-2-methylbutan-2-yl)carbamate hydrochloride 97%

Supplier: Ambeed
Warning

A139925-1G A139925-250MG A139925-100MG
76972-540EA 141 USD
76972-540 76972-542 77881-096
tert-Butyl (4-amino-2-methylbutan-2-yl)carbamate hydrochloride 97%
tert-Butyl (4-amino-2-methylbutan-2-yl)carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₃ClN₂O₂
MDL Number: MFCD12755995
CAS Number: 1179359-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR