2-Indoleboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

A129724-100MG A129724-10G A129724-1G A129724-250MG A129724-25G A129724-5G
76972-002EA 16 USD
76972-002 76972-004 76972-006 76972-008 76972-010 77670-836
2-Indoleboronic acid pinacol ester 97%
2-Indoleboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₂
MDL Number: MFCD09751284
CAS Number: 476004-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR