tert-Butyl (1-(4-aminophenyl)cyclobutyl)carbamate 95%

Supplier: Ambeed
Warning

A191339-1G A191339-250MG A191339-25G A191339-5G
76974-860EA 13 USD
76974-860 76974-862 76974-864 76974-866
tert-Butyl (1-(4-aminophenyl)cyclobutyl)carbamate 95%
tert-Butyl (1-(4-aminophenyl)cyclobutyl)carbamate

Precaución: for Research Use Only

Formula: C₁₅H₂₂N₂O₂
MDL Number: MFCD18762015
CAS Number: 1259224-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR