1H-Tetrazole-5-acetic acid 95%

Supplier: Ambeed
Danger

A295044-1G A295044-25G
76977-636EA 11.67 USD
76977-636 76977-638
1H-Tetrazole-5-acetic acid 95%
1H-Tetrazole-5-acetic acid

Precaución: for Research Use Only

Formula: C₃H₄N₄O₂
MW: 128.09 g/mol
MDL Number: MFCD03094010
CAS Number: 21743-75-9
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR