Methyl 4-bromo-5-cyano-3-methylthiophene-2-carboxylate 95%

Supplier: Ambeed
Warning

A685339-100MG A685339-1G A685339-250MG
76981-022EA 14.86 USD
76981-022 76981-024 76981-026
Methyl 4-bromo-5-cyano-3-methylthiophene-2-carboxylate 95%
Methyl 4-bromo-5-cyano-3-methylthiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₆BrNO₂S
MW: 260.11 g/mol
MDL Number: MFCD19703224
CAS Number: 648412-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR