5,7-Difluoroquinoline 95%

Supplier: Ambeed
Warning

A542668-1G A542668-250MG
76980-192EA 88.06 USD
76980-192 76980-194
5,7-Difluoroquinoline 95%
5,7-Difluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₅F₂N
MW: 165.1400 g/mol
MDL Number: MFCD00474638
CAS Number: 34522-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR