6-Bromo-7-fluoroquinoline 98%

Supplier: Ambeed
Warning

A133984-1G A133984-250MG A133984-5G A133984-25G
76972-252EA 22 USD
76972-252 76972-254 77566-614 77670-916
6-Bromo-7-fluoroquinoline 98%
6-Bromo-7-fluoroquinoline

Precaución: For research use only.

Formula: C₉H₅BrFN
MDL Number: MFCD13181511
CAS Number: 127827-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR