5-Amino-2-bromoisonicotinic acid 96%

Supplier: Ambeed
Warning

A169677-100MG A169677-1G A169677-250MG A169677-5G
76973-934EA 23.34 USD
76973-934 76973-936 76973-938 76973-940
5-Amino-2-bromoisonicotinic acid 96%
5-Amino-2-bromoisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
MDL Number: MFCD17016120
CAS Number: 1242336-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR