N-(2,3-Dimethyl-5,6,7,8-tetrahydrofuro[2,3-b]quinolin-4-yl)-2-(2-oxopyrrolidin-1-yl)acetamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Coluracetam
A103396-10MG
A103396-1MG
A103396-50MG
A103396-5MG
A103396-250MG
A103396-1G
A103396-100MG
76970-274EA
8.49
USD
76970-274
76970-276
76970-278
76970-280
77610-782
77610-780
77670-498
N-(2,3-Dimethyl-5,6,7,8-tetrahydrofuro[2,3-b]quinolin-4-yl)-2-(2-oxopyrrolidin-1-yl)acetamide 98%
N-(2,3-Dimethyl-5,6,7,8-tetrahydrofuro[2,3-b]quinolin-4-yl)-2-(2-oxopyrrolidin-1-yl)acetamide
Precaución: for Research Use Only
Formula:
C₁₉H₂₃N₃O₃ MW: 341.40 g/mol |
MDL Number:
MFCD00901416 CAS Number: 135463-81-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...