2-Fluoro-4-(trifluoromethyl)phenylboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-trifluoromethylphenylboronic acid pinacol ester, 2-(2-Fluoro-4-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester

A348452-1G A348452-250MG A348452-5G
76978-492EA 25 USD
76978-492 76978-494 76978-496
2-Fluoro-4-(trifluoromethyl)phenylboronic acid pinacol ester 95%
2-Fluoro-4-(trifluoromethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BF₄O₂
MDL Number: MFCD08458187
CAS Number: 1073353-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR