4-(Trifluoromethyl)pyrimidin-2(1H)-one 97%
Supplier: Ambeed
|
Warning
|
A240128-100G
A240128-10G
A240128-25G
A240128-500G
A240128-5G
76976-426EA
120.95
USD
76976-426
76976-428
76976-430
76976-432
76976-434
4-(Trifluoromethyl)pyrimidin-2(1H)-one 97%
4-(Trifluoromethyl)pyrimidin-2(1H)-one
Precaución: for Research Use Only
Formula:
C₅H₃F₃N₂O MW: 164.09 g/mol |
MDL Number:
MFCD00115016 CAS Number: 104048-92-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...