Z-Asp-OMe 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl N-Benzyloxycarbonyl-L-aspartate, N-Benzyloxycarbonyl-L-aspartic acid 1-Methyl ester, Cbz-Asp-OMe, 1-Methyl N-Cbz-L-aspartate, Z-Asp-OMe, N-Cbz-L-aspartic acid 1-Methyl ester, N-Benzyloxycarbonyl-L-aspartic acid 1-methyl ester, (S)-3-(((Benzyloxy)carbonyl)amino)-4-methoxy-4-oxobutanoic acid, 1-Methyl N-Carbobenzoxy-L-aspartate, N-Carbobenzoxy-L-aspartic acid 1-Methyl ester

A171264-1G A171264-100G A171264-25G A171264-5G A171264-500G
76974-012EA 29.71 USD
76974-012 76974-014 77671-434 77671-436 77671-438 77881-319
Z-Asp-OMe 98%
Z-Asp-OMe

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₆
MDL Number: MFCD00083277
CAS Number: 4668-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR