4,4'-Disulfanediylbis(2-aminobutanoic acid) 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-[(3-amino-4-hydroxy-4-oxo-butyl)disulphanyl]butanoic acid

A371556-100G A371556-10G A371556-25G A371556-500G A371556-5G
76978-702EA 132.62 USD
76978-702 76978-704 76978-706 76978-708 76978-710
4,4'-Disulfanediylbis(2-aminobutanoic acid) 95%
4,4'-Disulfanediylbis(2-aminobutanoic acid)

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₄S₂
MW: 268.3500 g/mol
MDL Number: MFCD00063095
CAS Number: 462-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR