5-Methyl-3-(trifluoromethyl)-1H-pyrazole 95+%

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-3-(trifluoromethyl)pyrazole

A708776-100MG A708776-1G A708776-250MG
76981-142EA 29.71 USD
76981-142 76981-144 76981-146
5-Methyl-3-(trifluoromethyl)-1H-pyrazole 95+%
5-Methyl-3-(trifluoromethyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₅H₅F₃N₂
MW: 150.10 g/mol
MDL Number: MFCD00156058
CAS Number: 942060-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR