4-Bromo-1H-pyrazole-5-carboxamide 95%

Supplier: Ambeed
Warning

A226996-250MG A226996-5G
76975-982EA 16 USD
76975-982 77881-579
4-Bromo-1H-pyrazole-5-carboxamide 95%
4-Bromo-1H-pyrazole-5-carboxamide

Precaución: for Research Use Only

Formula: C₄H₄BrN₃O
MDL Number: MFCD15146440
CAS Number: 1146951-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR