1-(1-Boc-piperidino)-1H-pyrazole-4-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: Crizotinib Impurity 5, 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]piperidine-1-carboxylic acid tert-Butyl ester, tert-Butyl 4-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)piperidine-1-carboxylate, 1-(1-Boc-piperidin-4-yl)pyrazole-4-boronic acid Pinacol ester, 2-[1-(1-Boc-piperidin-4-yl)pyrazol-4-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, tert-Butyl 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]piperidine-1-carboxylate

A153059-100G A153059-10G A153059-250MG A153059-25G A153059-500G A153059-5G A153059-1G A153059-100MG
76973-176EA 167 USD
76973-176 76973-178 76973-180 76973-182 76973-184 76973-186 77671-144 77881-179
1-(1-Boc-piperidino)-1H-pyrazole-4-boronic acid pinacol ester 98%
1-(1-Boc-piperidino)-1H-pyrazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₃₂BN₃O₄
MDL Number: MFCD11112131
CAS Number: 877399-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR