tert-Butyl 5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazine-7(8H)-carboxylate 97%

Supplier: Ambeed
Warning

A208098-100MG A208098-1G
76975-416EA 36.08 USD
76975-416 77612-044
tert-Butyl 5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazine-7(8H)-carboxylate 97%
tert-Butyl 5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazine-7(8H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₄O₂
MW: 224.26 g/mol
MDL Number: MFCD10574958
CAS Number: 723286-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR