2-Chloropropyldimethylammonium chloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(2-Chloropropyl)-N,N-dimethylammonium chloride, 2-Chloro-N,N-dimethylpropan-1-amine hydrochloride, 2-Dimethylaminoisopropyl chloride hydrochloride
A225095-1000G
A225095-100G
A225095-25G
A225095-500G
76975-964EA
195.21
USD
76975-964
76975-966
76975-968
76975-970
2-Chloropropyldimethylammonium chloride 98%
2-Chloropropyldimethylammonium chloride
Precaución: for Research Use Only
Formula:
C₅H₁₃Cl₂N MW: 158.07 g/mol |
MDL Number:
MFCD00012534 CAS Number: 4584-49-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...