4-Cyclopropylphenylboronic acid 97%

Supplier: Ambeed
Warning

A208820-100MG A208820-1G A208820-250MG A208820-5G
76975-454EA 18 USD
76975-454 76975-456 76975-458 76975-460
4-Cyclopropylphenylboronic acid 97%
4-Cyclopropylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₂
MDL Number: MFCD01309064
CAS Number: 302333-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR