6-Acetyl-1H-indazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(1H-Indazol-6-yl)ethan-1-one, 1-(1H-Indazol-6-yl)ethanone
A718225-1G
A718225-250MG
76981-226EA
388.29
USD
76981-226
76981-228
6-Acetyl-1H-indazole 97%
6-Acetyl-1H-indazole
Precaución: for Research Use Only
Formula:
C₉H₈N₂O MW: 160.173 g/mol |
MDL Number:
MFCD11869764 CAS Number: 189559-85-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...