1,3-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,3-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1,3-Dimethylpyrazole-4-boronic acid Pinacol ester, 1,3-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 2-(1,3-Dimethyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
A390521-100G
A390521-10G
A390521-1G
A390521-250MG
A390521-25G
A390521-5G
76978-932EA
257
USD
76978-932
76978-934
76978-936
76978-938
76978-940
76978-942
1,3-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester 97%
1,3-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₁H₁₉BN₂O₂ |
MDL Number:
MFCD09864190 CAS Number: 1046832-21-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...