tert-Butyl-4,5-dihydro-1H-pyrazolo[3,4-c]pyridine-6(7H)-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4,5-dihydro-1H-pyrazolo[3,4-c]pyridine-6(7H)-carboxylate

A160555-100MG A160555-250MG A160555-1G
76973-570EA 43.5 USD
76973-570 76973-572 77550-016
tert-Butyl-4,5-dihydro-1H-pyrazolo[3,4-c]pyridine-6(7H)-carboxylate 97%
tert-Butyl-4,5-dihydro-1H-pyrazolo[3,4-c]pyridine-6(7H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₇N₃O₂
MW: 223.27 g/mol
MDL Number: MFCD11044813
CAS Number: 871726-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR