1-Bromo-3-chloro-2,5-difluorobenzene 98%

Supplier: Ambeed
Warning

A397182-1G A397182-250MG A397182-5G
76979-010EA 35 USD
76979-010 76979-012 76979-014
1-Bromo-3-chloro-2,5-difluorobenzene 98%
1-Bromo-3-chloro-2,5-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClF₂
MDL Number: MFCD11845991
CAS Number: 1160573-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR