α,α,α-Trifluoro-3-tolualdehyde 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Trifluoromethyl)benzaldehyde, Cinacalcet Impurity 23

A110535-1000G A110535-100G A110535-10G A110535-25G A110535-5G A110535-500G
76970-724EA 189.91 USD
76970-724 76970-726 76970-728 76970-730 76970-732 77670-568
α,α,α-Trifluoro-3-tolualdehyde 97%
α,α,α-Trifluoro-3-tolualdehyde

Precaución: for Research Use Only

Formula: C₈H₅F₃O
MW: 174.12 g/mol
MDL Number: MFCD00003373
CAS Number: 454-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR