2-Butyl-5-nitrobenzofuran 97%

Supplier: Ambeed
Warning

A316866-100MG A316866-10G A316866-250MG A316866-5G A316866-100G A316866-1G
76977-994EA 6 USD
76977-994 76977-996 76977-998 76978-000 77672-620 77672-622
2-Butyl-5-nitrobenzofuran 97%
2-Butyl-5-nitrobenzofuran

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₃
MDL Number: MFCD14525624
CAS Number: 133238-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR