3-(Chloromethyl)-5-methylpyridine hydrochloride 97%

Supplier: Ambeed
Danger

A227116-100MG A227116-1G A227116-5G A227116-10G A227116-250MG A227116-25G
76975-984EA 5.31 USD
76975-984 76975-986 77593-468 77612-210 77671-998 77672-000
3-(Chloromethyl)-5-methylpyridine hydrochloride 97%
3-(Chloromethyl)-5-methylpyridine hydrochloride

Precaución: for Research Use Only

Formula: C7H9Cl2N
MW: 178.0600 g/mol
MDL Number: MFCD11656303
CAS Number: 1007089-84-0
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR