4-Borono-3-fluorobenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Carboxy-2-fluorophenylboronic acid

A125374-1G A125374-250MG A125374-25G A125374-5G
76971-826EA 15 USD
76971-826 76971-828 76971-830 76971-832
4-Borono-3-fluorobenzoic acid 97%
4-Carboxy-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BFO₄
MDL Number: MFCD08458473
CAS Number: 851335-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR