6-Chloronicotinamidine hydrochloride 95%

Supplier: Ambeed
Warning

A103621-100MG A103621-250MG
76970-288EA 8 USD
76970-288 76970-290
6-Chloronicotinamidine hydrochloride 95%
6-Chloronicotinamidine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₇Cl₂N₃
MDL Number: MFCD11052371
CAS Number: 201937-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR