Tris(phenylthio)methane 98%

Supplier: Ambeed
Warning

A674024-1G A674024-25G A674024-5G
76980-930EA 7.43 USD
76980-930 76980-932 76980-934
Tris(phenylthio)methane 98%
Tris(phenylthio)methane

Precaución: for Research Use Only

Formula: C₁₉H₁₆S₃
MW: 340.52 g/mol
MDL Number: MFCD00010396
CAS Number: 4832-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR