N-Boc-glycine (boc-gly-OH) 97%

Supplier: Ambeed
Danger

Synonyms: Boc-Gly-OH

A133228-100G A133228-10G A133228-25G A133228-500G A133228-1000G
76972-202EA 20 USD
76972-202 76972-204 76972-206 76972-208 77670-896
N-Boc-glycine (boc-gly-OH) 97%
N-Boc-glycine (Boc-Gly-OH)

Precaución: for Research Use Only

Formula: C₇H₁₃NO₄
MW: 175.2 g/mol
MDL Number: MFCD00002690
CAS Number: 4530-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR