4-Methylpyrocatechol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methylbenzene-1,2-diol, 4-Methylcatechol

A111032-100G A111032-10G A111032-25G A111032-500G A111032-5G
76970-782EA 32.89 USD
76970-782 76970-784 76970-786 76970-788 76970-790
4-Methylpyrocatechol 98%
4-Methylpyrocatechol

Precaución: for Research Use Only

Formula: C₇H₈O₂
MW: 124.14 g/mol
MDL Number: MFCD00002205
CAS Number: 452-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR