1-(4-Methoxypyridin-2-yl)ethanone 97%
Supplier: Ambeed
|
Warning
|
A623052-100MG
A623052-1G
A623052-5G
76980-638EA
46.68
USD
76980-638
76980-640
76980-642
1-(4-Methoxypyridin-2-yl)ethanone 97%
1-(4-Methoxypyridin-2-yl)ethanone
Precaución: for Research Use Only
Formula:
C₈H₉NO₂ |
MDL Number:
MFCD10697664 CAS Number: 59576-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...