5-Chloro-2-fluorotoluene 97%

Supplier: Ambeed
Danger

A788566-25G A788566-5G
76981-596EA 13 USD
76981-596 76981-598
5-Chloro-2-fluorotoluene 97%
5-Chloro-2-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₆ClF
MDL Number: MFCD00018504
CAS Number: 452-66-4
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR