Brivanib (BMS-540215) 96%

Supplier: Ambeed
Warning

Synonyms: (R)-1-((4-((4-Fluoro-2-methyl-1H-indol-5-yl)oxy)-5-methylpyrrolo[2,1-f][1,2,4]triazin-6-yl)oxy)propan-2-ol, Brivanib

A699752-10MG A699752-1MG A699752-2MG A699752-5MG A699752-100MG A699752-250MG A699752-50MG
76981-078EA 45.62 USD
76981-078 76981-080 76981-082 76981-084 77614-168 77673-910 77673-912
Brivanib (BMS-540215) 96%
Brivanib (BMS-540215)

Precaución: for Research Use Only

Formula: C₁₉H₁₉FN₄O₃
MW: 370.4 g/mol
MDL Number: MFCD13194684
CAS Number: 649735-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR