3-Bromo-4-fluorotoluene 98%

Supplier: Ambeed
Warning

A300622-100G A300622-25G A300622-5G
76977-718EA 39 USD
76977-718 76977-720 76977-722
3-Bromo-4-fluorotoluene 98%
3-Bromo-4-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₆BrF
MDL Number: MFCD00040827
CAS Number: 452-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR