3-(2-Nitrophenyl)-1H-pyrazole 95%

Supplier: Ambeed
Warning

A207769-100MG A207769-1G A207769-250MG
76975-404EA 33.95 USD
76975-404 76975-406 76975-408
3-(2-Nitrophenyl)-1H-pyrazole 95%
3-(2-Nitrophenyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₇N₃O₂
MDL Number: MFCD00665858
CAS Number: 59844-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR