Boc-Cit-OH 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-5-ureidopentanoic acid

A614820-1G A614820-25G A614820-5G
76980-574EA 28.65 USD
76980-574 76980-576 76980-578
Boc-Cit-OH 95%
Boc-Cit-OH

Precaución: for Research Use Only

Formula: C₁₁H₂₁N₃O₅
MW: 275.305 g/mol
MDL Number: MFCD00080271
CAS Number: 45234-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR